Cleanroom gloves, polychloroprene, BioClean Fusion™
Dodavatel: Nitritex
Certifikáty
O tomto produktu
These ambidextrous gloves with beaded cuffs provide excellent grip. The non sterile version is suitable for chemotherapy applications (ASTM D 6978-05 tested).
- Sterile or non sterile, powder-free, textured finish
- Latex-free to eliminate Type 1 latex allergies
- Beaded cuff provides easier donning and stability on the arm
- Polychloroprene provides exceptional tactility
- Suitable for double donning
- Excellent ESD properties
Suitable for use in Class 10, ISO 4 or M2.5 cleanrooms.
EN 374-1/2/3, EN 455; Cat. III, AQL 1,5
Certifications: Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 +A1:2009, EN 374-1:2016, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Sterilized in accordance with ISO 11137-1:2006.
Suitable for contact with all foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Tested against permeation standard ASTM D6978-05 for cytotoxic drugs.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 +A1:2009, EN 374-1:2016, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Sterilized in accordance with ISO 11137-1:2006.
Suitable for contact with all foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Tested against permeation standard ASTM D6978-05 for cytotoxic drugs.
Packaging: Sterile version: One cuffed pair per plastic wallet; one wallet per EasyTear™ PE pouch; 10 pairs per sealed outer PE bag; 20 outer bags per carton liner; one carton liner per carton (200 pairs). Non sterile version: 100 pieces per sealed inner PE bag; one sealed inner PE bag per sealed outer PE bag; 10 outer PE bags per carton liner; one carton liner per carton (1000 pieces).
Specifikace
- Catalog No:
- 112-3899
- 112-3838
- 112-3896
- 112-3895
- 112-3894
- 112-3834
- 112-3499
- 112-3836
- 112-3837
- 112-3898
- 112-3835
- 112-3897
- Model:
- Sterile
- Non sterile
- Sterile
- Sterile
- Sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Sterile
- Non sterile
- Sterile
- Obouruké/jednoruké:
- Obouruké
- Obouruké
- Obouruké
- Obouruké
- Obouruké
- Obouruké
- Obouruké
- Obouruké
- Obouruké
- Obouruké
- Obouruké
- Obouruké
- Jednorázové/znovupoužitelné:
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Latex/bez latexu:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Pudrované/nepudrované:
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Třída čistoty:
- 10/ISO 4; EU GMP (A)
- 10/ISO 4
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4; EU GMP (A)
- 10/ISO 4
- 10/ISO 4; EU GMP (A)
- Typ manžety:
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Tloušťka límce:
- 0,07 mm
- 0,07 mm
- 0,07 mm
- 0,07 mm
- 0,07 mm
- 0,07 mm
- 0,07 mm
- 0,07 mm
- 0,07 mm
- 0,07 mm
- 0,07 mm
- 0,07 mm
- Tloušťka prstu:
- 0,12 mm
- 0,12 mm
- 0,12 mm
- 0,12 mm
- 0,12 mm
- 0,12 mm
- 0,12 mm
- 0,12 mm
- 0,12 mm
- 0,12 mm
- 0,12 mm
- 0,12 mm
- Tloušťka drapáku:
- 0,10 mm
- 0,10 mm
- 0,10 mm
- 0,10 mm
- 0,10 mm
- 0,10 mm
- 0,10 mm
- 0,10 mm
- 0,10 mm
- 0,10 mm
- 0,10 mm
- 0,10 mm
- Norma EN:
- EN 374-1/2/3, EN 455
- EN 374-1/2/3, EN 455
- EN 374-1/2/3, EN 455
- EN 374-1/2/3, EN 455
- EN 374-1/2/3, EN 455
- EN 374-1/2/3, EN 455
- EN 374-1/2/3, EN 455
- EN 374-1/2/3, EN 455
- EN 374-1/2/3, EN 455
- EN 374-1/2/3, EN 455
- EN 374-1/2/3, EN 455
- EN 374-1/2/3, EN 455
Specifikace
Související informace
Frequently Bought Together