Carrez II solution zinc acetate 1 mol/l in acetic acid solution (0,5 mol/l)
Catalog # 5056.1000
Supplier: VWR Chemicals
CAS Number:
Specifications
- Typ balení:Glass bottle
- Bal.:1 L
- Sekvence:5037
Specifications
About this item
:
Storage Temperature:Ambient
UN:3082
ADR:9,III
:GHS09