Test strips, lactic acid, Reflectoquant®, Supelco®
Catalog # 1.16127.0001
Supplier: Merck
CAS Number:
Specifications
- Chemická metoda:Enzymatic reaction
- Bal.:50 Tests
- Popis:Lactic acid test
- Rozsah:3 - 60,0 mg/l CH₃CHOHCO₂H
Specifications
About this item
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
The test measures total lactic acid (sum of D- and L-lactic acid in beverages, e.g. wine, beer and fruit/vegetable juice after dilution. Furthermore the test can be used for the determination of lactic acid in food after pretreatment (typically dilution).