Reflectoquant®10 plus Test kits, Supelco®
1.17945.0001
: Merck
:
- Chemická metoda:Kalignost
- Bal.:100 Tests
- Popis:Reflectoquant® 10 plus Potassium test
- M-Clarity quality level:MQ100
- Rozsah:1,0 - 25,0 mg/l K
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly