Test strips, total sugar (glucose and fructose), Reflectoquant®, Supelco®
Catalog # 1.16136.0001
Supplier: Merck
CAS Number:
Specifications
- Chemická metoda:Enzymatic reaction
- Bal.:50 Tests
- Popis:Total sugar test (glucose and fructose)
- M-Clarity quality level:MQ100
- Rozsah:65 - 650 mg/l
Specifications
About this item
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly