Test strips, formaldehyde, Reflectoquant®, Supelco®
Catalog # 1.16989.0001
Supplier: Merck
CAS Number:
Specifications
- Chemická metoda:Triazole
- Bal.:50 Tests
- Popis:Formaldehyde test
- M-Clarity quality level:MQ100
- Rozsah:1,0 - 45,0 mg/l HCHO
Specifications
About this item
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly