Reagent Kits for Protein and Fibre Analysis, BIOQUANT®, Supelco®
Catalog # 1.10537.0001
Supplier: Merck
CAS Number:
Specifications
- Chemická metoda:Carrez Clarification
- Bal.:1 KIT
- Popis:Reagent kit for sample preparation in food analysis, 5-fold concentrate
- M-Clarity quality level:MQ100