Orders LinkContactUsLinkComponent
QUANTOFIX® Test Strips, Total Iron
QUANTOFIX® Test Strips, Total Iron
18, QUANTOFIX® tin, 0 • 10 • 25 • 50 • 100 • 250 • 500 - mg/L - Sn²⁺, Color reaction with molybdenum phosphoric acid, 40×40×120 mm (packaging), 100 Tests, 100 Tests
Catalog # 705-0310
Supplier:   MACHEREY-NAGEL
QUANTOFIX® Test Strips, Total Iron
Catalog # 705-0310
Supplier:  MACHEREY-NAGEL
CAS Number:  
QUANTOFIX® Test Strips, Total Iron
QUANTOFIX® Test Strips, Total Iron
18, QUANTOFIX® tin, 0 • 10 • 25 • 50 • 100 • 250 • 500 - mg/L - Sn²⁺, Color reaction with molybdenum phosphoric acid, 40×40×120 mm (packaging), 100 Tests, 100 Tests
CAS Number:  
Price
1 460,00 Kč
Log in for availability
1 * 100 Tests
Product Details & Documents
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips allow the quick and easy determination of total iron in solutions. The easy dip-and-read-procedure provides reliable results without the need for training or experience. Within one minute one gets reliable results.
Specifications
Specifications
  • Chemická metoda:
    Color reaction with molybdenum phosphoric acid
  • Bal.:
    100 Tests
  • Popis:
    QUANTOFIX® tin
  • Rozměry:
    40×40×120 mm (packaging)
  • Sekvence:
    18
  • Testů v kitu:
    100 Tests
  • Rozsah:
    0 • 10 • 25 • 50 • 100 • 250 • 500 - mg/L - Sn²⁺
  • Obsah:
    100 test strips in a tube, instruction leaflet
  • Hmotnost:
    42,1 g
  • Cat. No.:
    705-0310
  • Plošina:
    Semi-quantitative test strips
  • Parametry:
    All reagents included
  • Přístroj:
    No
  • Životnost police (měsíce):
    30 (from production)
  • Shoda:
    No hazardous material
  • Metoda:
    Tin - Phosphomolybdic acid (white → dark blue)