Orders LinkContactUsLinkComponent
Test strips, nitrite, MQuant®, Supelco®
Test strips, nitrite, MQuant®, Supelco®
Dodavatel:   Merck
Test strips, nitrite, MQuant®, Supelco®
Dodavatel:  Merck
Certifikáty
Test strips, nitrite, MQuant®, Supelco®
Test strips, nitrite, MQuant®, Supelco®
3 Dostupných variant níže
Product Details & Documents
MQuant® (MQ) test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically-degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
  • Easy to handle: just dip, wait and measure visually against colour scale
  • Safe: no auxiliary reagents necessary
  • Immediate results within 2 minutes enables fast decisions
  • Portable: can be used inside or outside the laboratory
Specifications for Product Family
Specifikace
Specifikace
  • Catalog No:
  • 1.10022.0001
  • 1.10057.0001
  • 1.10007.0002
  • M-Clarity quality level:
  • MQ100
  • MQ100
  • MQ100
Specifikace
Product Family Options
Informace o produktu
  • Sekvence
    Rozsah
    Chemická metoda
    Typ
    Bal.
    Dostupnost
    Price
  • Nitrite test
    01
    0,1-0,3-0,6-1-2-3 g/l NO₂
    Griess' reaction
    MQ
    100 Tests
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 100 Tests
    1 * 100 Tests - 1 250,00 Kč
    1 250,00 Kč
    1 * 100 Tests
       1 250,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.10022.0001
    M-Clarity quality level
    MQ100
  • Nitrite test
    01
    0-0,5-1-2-5-10 mg/l NO₂
    Griess' reaction
    MQ
    75 Tests
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 75 Tests
    1 * 75 Tests - 1 110,00 Kč
    1 110,00 Kč
    1 * 75 Tests
       1 110,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.10057.0001
    M-Clarity quality level
    MQ100
  • Nitrite test
    03
    2-5-10-20-40-80 mg/l NO₂
    Griess' reaction
    MQ
    25 Tests
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 25 Tests
    1 * 25 Tests - 772,00 Kč
    772,00 Kč
    1 * 25 Tests
       772,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.10007.0002
    M-Clarity quality level
    MQ100