Orders LinkContactUsLinkComponent
Test Strips, nitrate, MQuant®, Supelco®
Test Strips, nitrate, MQuant®, Supelco®
Dodavatel:   Merck
Test Strips, nitrate, MQuant®, Supelco®
Dodavatel:  Merck
Certifikáty
Test Strips, nitrate, MQuant®, Supelco®
3 Dostupných variant níže
Product Details & Documents
MQuant® (MQ) test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically-degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
  • Easy to handle: just dip, wait and measure visually against colour scale
  • Safe: no auxiliary reagents necessary
  • Immediate results within 2 minutes enables fast decisions
  • Portable: can be used inside or outside the laboratory
Specifications for Product Family
Specifikace
Specifikace
  • Catalog No:
  • 1.10020.0001
  • 1.10020.0002
  • 1.10092.0021
  • M-Clarity quality level:
  • MQ100
  • MQ100
  • MQ100
Specifikace
Product Family Options
Informace o produktu
  • Rozsah
    Chemická metoda
    Typ
    Bal.
    Dostupnost
    Price
  • Nitrate test
    10-25-50-100-250-500 mg/l NO₃
    Griess' reaction (modified)
    MQ
    100 Tests
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 100 Tests
    1 * 100 Tests - 1 220,00 Kč
    1 220,00 Kč
    1 * 100 Tests
       1 220,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.10020.0001
    M-Clarity quality level
    MQ100
  • Nitrate test
    10-25-50-100-250-500 mg/l NO₃
    Griess' reaction (modified)
    MQ
    25 Tests
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 25 Tests
    1 * 25 Tests - 725,00 Kč
    725,00 Kč
    1 * 25 Tests
       725,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.10020.0002
    M-Clarity quality level
    MQ100
  • 1.10092.0021
    Nitrate test ( individually sealed)
    Nitrate test ( individually sealed)
    10-25-50-100-250-500 mg/l NO₃
    Griess' reaction (modified)
    MQ
    1.000 Tests
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1.000 Tests
    1 * 1.000 Tests - 12 710,00 Kč
    12 710,00 Kč
    1 * 1.000 Tests
       12 710,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1.10092.0021
    M-Clarity quality level
    MQ100