Test strips for cobalt, MQuant®, Supelco®
Catalog # 1.10002.0001
Supplier: Merck
CAS Number:
Specifications
- Chemická metoda:Rhodanide
- Bal.:100 Tests
- Popis:Cobalt test
- M-Clarity quality level:MQ100
- Typ:MQ
- Rozsah:10-30-100-300-1000 mg/l Co
Specifications
About this item
MQuant® (MQ) test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically-degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
- Easy to handle: just dip, wait and measure visually against colour scale
- Safe: no auxiliary reagents necessary
- Immediate results within 2 minutes enables fast decisions
- Portable: can be used inside or outside the laboratory