ContactUsLinkComponent
Visual colour kits, residual hardness, MQuant®, Supelco®
Visual colour kits, residual hardness, MQuant®, Supelco®
  1.11142.0001
 :  Merck
 :  
Visual colour kits, residual hardness, MQuant®, Supelco®
  1.11142.0001
 :  Merck
 :  
  • Chemická metoda:
    Mixed indicator
  • Bal.:
    400 Tests
  • Popis:
    Residual hardness test
  • M-Clarity quality level:
    MQ100
  • Typ:
    MTC
  • Rozsah:
    0,05 - 0,10 - 0,19 °e (0,7 - 1,4 - 2,7 mg/l CaCO₃) 0,07 - 0,14 - 0,27 °f
MQuant® titrimetric and colorimetric methods.
  • Results in approx. 1 minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available
 : Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.