Burette and test tube clamps, with bosshead
Katalogové číslo BOCH5631
Dodavatel: BOCHEM
Specifikace
- Určeno pro:Burettes and test tubes
- Upínací šířka:7 - 25 mm
- Bal.:1
Specifikace
O tomto produktu
These chromium plated clamps with bosshead (13,5 mm) and PVC coated jaws are designed for holding burettes and test tubes (Ø 7 - 25 mm).