Silikagel, Pore size: ∼60 Å 70-230 mesh, high purity for column chromatography, Supelco®
Catalog # 60740-500G
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:500 g
- Sekvence:3645
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
CAS Number:112926-00-8
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232