Douche cans/irrigators
02, 2 l, Douche can with flat back and suspension tag
Katalogové číslo KART590
Dodavatel: KARTELL
Přejít na:
Product Details & Documents
Douche cans/irrigators, made of polypropylene.
- Translucent
- With flat back and suspension tag
- Autoclavable
Specifikace
- Type:Douche can with flat back and suspension tag
- Kapacita:2 l
- Sekvence:02