Orders LinkContactUsLinkComponent
Spoon spatulas
Spoon spatulas
Supplier:   RSG
Spoon spatulas
Supplier:  RSG
Certificates
Spoon spatulas
4 Options Available Below
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spoon spatula for a wide range of uses.
  • With round handle
  • Very good thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Product Family Options
Product Information
  • Délka
    Availability
    Price
  • 150 mm
    Log in for availability
    Log in for availability
    1 * 5 KS
    1 * 5 KS - 1 940,00 Kč
    1 940,00 Kč
    1 * 5 KS
       1 940,00 Kč
  • 180 mm
    Log in for availability
    Log in for availability
    1 * 5 KS
    1 * 5 KS - 2 040,00 Kč
    2 040,00 Kč
    1 * 5 KS
       2 040,00 Kč
  • 210 mm
    Log in for availability
    Log in for availability
    1 * 5 KS
    1 * 5 KS - 2 220,00 Kč
    2 220,00 Kč
    1 * 5 KS
       2 220,00 Kč
  • 250 mm
    Log in for availability
    Log in for availability
    1 * 5 KS
    1 * 5 KS - 2 330,00 Kč
    2 330,00 Kč
    1 * 5 KS
       2 330,00 Kč