ContactUsLinkComponent
Beakers, low form, BOLA
Beakers, low form, BOLA
Catalog # BOHLA136-03
Supplier:  Bohlender
Supplier #: A136-03
undefined
Beakers, low form, BOLA
Catalog # BOHLA136-03
Supplier:  Bohlender
Supplier #: A136-03
Log in for availability
Select an Option
21 mm
Select an Option
5 ml
Select an Option
Select an Option
24 mm
Select an Option
02
Select an Option
1
About this item
PTFE, with spout.
  • Excellent chemical resistance
  • Temperature resistant from –200 to +250 °C
  • Flat bottom for good heat transfer, for example when used on regulated hotplates
Thick walled, with reinforced lip and exceptionally smooth inner walls to prevent contamination. Non stick. Low gas permeability.
Frequently Bought Together