Bis(2-ethylhexyl) adipate 2,000 µg/ml in methanol, Supelco®
47995-U
: Merck
:
- Koncentrace:2000 μg/ml in methanol
- Bal.:1
- Standard:Certified reference material
- Čí. CAS:103-23-1
- Popis:Bis(2-ethylhexyl) adipate 2,000 µg/ml in methanol
- Podmínky uskladnění:2…8 °C
- Popis rizikové třídy:H225 - H301 + H311 + H331 - H370
- Podmínky k odeslání:STD
- M-Clarity quality level:MQ100
- Synonym(s):Bis(2-ethylhexyl) adipate; DEHA
- GHS signal: GHS06, GHS08
: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311