Chlorocyclohexane for synthesis, Sigma-Aldrich®
: Merck
:542-18-7
:
:C₆H₁₁Cl
:MFCD00003822
:13,02761
:142 °C (1013 hPa)
:–44 °C
:1 g/cm³ (20 °C)
:Ambient
:208-806-6
:3,III
:M-Clarity™ quality level = MQ200
:1993
:GHS02
:GHS09
- Solubility:0.40 g/l (20 °C)
- Melting Point:-44 °C
- Molar Mass:118.6 g/mol
- Boiling Point:141 - 143 °C (1013 hPa)
- Vapor Pressure:7 hPa (20 °C)
- Flash Point:34 °C
- Refractive Index:1.4626 (20 °C, 589 nm)
- Explosion Limit:1.1 - 7.5 %(V)
- Density:1.00 g/cm3 (20 °C)
- Ignition Point:290 °C
Frequently Bought Together