Luminescence microplates, 96-well
Catalog # RATI6018710
Supplier: RATIOLAB
Specifications
- Poč. jamek:96
- Sterilní:+
- Barva:Černá
- Bal.:100
- Objem jamky:300 μl
Specifications
About this item
These wells are with raised rims for reduced cross-contamination and effective sealing with cover films. Precise design, perfectly shaped for use with automatic systems.
- SBS standard micro test plate format
- 8×12 wells alphanumerically coded
- Optimum space for precise positioning
- Space-saving and stackable
- Surface untreated