Inoculating loops and needles
Catalog # RATI7020200
Supplier: RATIOLAB
Specifications
- Balené:100 peel packs of 20
- Kapacita:10 µl
- Popis:Loop
- Bal.:2.000
Specifications
About this item
Made of polystyrene (PS).
- Hexagonal handle for safe handling
- Beta-sterile
- Sterile, individually packed in easy to open peel-packs