Stopcocks for burettes, angled, DURAN®
2,0 mm, 12,5 NS
Katalogové číslo SCOT286451205
Dodavatel: DWK Life Sciences
Přejít na:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, fine ground, without keys.
- Very good chemical resistance
- Very good temperature resistance
For glass manipulators.
Specifikace
- Ø otvoru:2,0 mm
- Kohout:12,5 NS