2D data matrix bar code bottom inserts, WHEATON®
2D data matrix bar code bottom inserts, non sterile
Katalogové číslo 479-0918
Dodavatel: DWK Life Sciences
2D data matrix bar code bottom inserts, WHEATON®
Katalogové číslo 479-0918
Dodavatel: DWK Life Sciences
Přejít na:
Product Details & Documents
These 2D data matrix bar code inserts press fit and lock into place in the bottom of the vial. They are made of the same PP as the vials themselves and are laser-etched for durability. They allow for immediate bar coding of samples. The insert can also be applied to the vial at a future date, which eliminates jeopardising the integrity of the sample by transferring it to another vial.
Documents
VWRBRO_007b Biobanking_EU
Produktová literatura
Specifikace
- Popis:2D data matrix bar code bottom inserts, non sterile