Stopcocks, NO-DRIP
"Zero drip" tap made of 18/10 stainless steel
Katalogové číslo 216-0235
Dodavatel: BOCHEM
Přejít na:
Product Details & Documents
This stopcock are already mounted, PTFE seal with max. 260 °C.
Specifikace
- Popis:"Zero drip" tap made of 18/10 stainless steel