Accessories for Orion™ AQUAfast turbidimeters
Frequently Bought Together
Masterflex® L/S® Precision Pump Tubing, Tygon® A-60-F and A-70-F, Avantor®
: Avantor Fluid Handling
VWR® AVS TITRINORM, pufrovací roztoky, 20 °C
: VWR Chemicals
Buffer solutions in sachets
: Mettler - Toledo
Roztoky standardní pro měření vodivosti
: VWR Chemicals
pH buffers, ready to use
: WTW