Orders LinkContactUsLinkComponent
Stand for Imhoff funnels
Stand for Imhoff funnels
for two Imhoff funnels
Katalogové číslo BURK7118-2000
Dodavatel:   BURKLE
Stand for Imhoff funnels
Katalogové číslo BURK7118-2000
Dodavatel:  BURKLE
Stand for Imhoff funnels
Stand for Imhoff funnels
for two Imhoff funnels
Price
2 930,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
PP, chrome-plated steel. The stand for sedimentation funnels is used to keep, hold or store Imhoff funnels safely and reliably during sedimentation.
  • Adjustable height up to 450 mm
  • For funnels with Ø 90 mm or more
Two funnel holders ensure that Imhoff funnels will be held safely and securely and can be placed in an absolutely vertical position. Thanks to an innovative magnet concept no clamping screws are necessary. The height of the funnel holder can be adjusted easily and flexibly with one hand. With the corresponding accessory the holders for Imhoff funnels can also be replaced by holders for all-purpose funnels.
Specifikace
  • Popis:
    for two Imhoff funnels
Accessories
Informace o produktu
  • Katalogové číslo
    Dostupnost
    Price
  • Držák pro nálevky s redukční vložkou, pro nálevky s Ø 40 - 180 mm
    Držák pro nálevky s redukční vložkou, pro nálevky s Ø 40 - 180 mm
    612-5698
    Přihlásit se pro dostupnost
    1 * 1 KS - 446,00 Kč
Informace o produktu
  • Katalogové číslo
    Dostupnost
    Price
  • Držák pro sedimentační válce dle Imhoffa s Ø 90 mm a více
    Držák pro sedimentační válce dle Imhoffa s Ø 90 mm a více
    612-5699
    Přihlásit se pro dostupnost
    1 * 1 KS - 563,00 Kč