Clips assortment for conical joints, POM, KECK™, DURAN®
SCOT290310006
- Pro NZ:10 / 14 / 19 / 24 / 29 / 34 / 45
- Barva:Různé
- Bal.:1
Polyoxymethylene, coloured.
: The set includes the following clips: 2 × KC 14, 2 × KC 19, 1 × KC 29, 1 × KC 10, 1 × KC 24, 1 × KC 34, 1 × KC 45.