Orders LinkContactUsLinkComponent
Forceps, precision, De Bakey
Forceps, precision, De Bakey
Precision forceps, De Bakey, Straight, 160 mm
Catalog # BOCH1960
Supplier:   BOCHEM
Forceps, precision, De Bakey
Catalog # BOCH1960
Supplier:  BOCHEM
Forceps, precision, De Bakey
Forceps, precision, De Bakey
Precision forceps, De Bakey, Straight, 160 mm
Price
3 610,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
'De Bakey' precision forceps made of titanium.
Specifications
  • Type:
    Straight
  • Popis:
    Precision forceps, De Bakey
  • Délka:
    160 mm