Stand for three pipettes
Catalog # KART1315
Supplier: KARTELL
Specifications
- Barva:Bezbarvá
- Popis:Stand for 3 pipettors, 145×70 mm
- Bal.:1
Specifications
About this item
Transparent plastic holder for three Kartell micropipettes (Pluripet, Pluripet II, Fixopet, Labo!x) or pipettes from other brands.
- Robust, easy to clean
- Great stability thanks to the wide base
- Sterilisable with radiation