Oxid hlinitý ≥99.8% (trace metals basis), nanopowder 13 nm (TEM), Sigma-Aldrich®
Catalog # SIAL718475-100G
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:100 g
- Sekvence:3401
Specifications
About this item
:
Merck Index:13,00355
Boiling Pt:2980 °C (1013 hPa)
Melting Pt:2045 °C
Density:3,94 g/cm³ (20 °C)
EINECS:215-691-6
REACH:01-2119529248-35
CAS Number:1344-28-1
:11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424