Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Zkumavky
Mikrozkumavky
Accessories for cluster tube racks, 1,2 ml, BioTube™
Accessories for cluster tube racks, 1,2 ml, BioTube™
SIMPT101-50
:
Simport Scientific
Accessories for cluster tube racks, 1,2 ml, BioTube™
SIMPT101-50
:
Simport Scientific
Popis:
Storage box only for BioTube™ rack, without tubes
Bal.:
10