Orders LinkContactUsLinkComponent
Separating funnels, conical, PMP
Separating funnels, conical, PMP
500 ml, 29/32 NS
Katalogové číslo KART978
Dodavatel:   KARTELL
Separating funnels, conical, PMP
Katalogové číslo KART978
Dodavatel:  KARTELL
Separating funnels, conical, PMP
Separating funnels, conical, PMP
500 ml, 29/32 NS
Přejít na:
Price
2 220,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Separating funnels, made of PMP, crystal clear, with permanent graduations.
  • Needle control for precise control of flow
  • Top unscrews and valve can be detached for thorough cleaning
  • Autoclavable and chemical resistant
  • Complete with stopper
Documents
Specifikace
  • Velikost objímky:
    29/32 NS
  • Kapacita:
    500 ml