Orders LinkContactUsLinkComponent
Wire baskets, rectangular, with handle
Wire baskets, rectangular, with handle
300×400×200 mm
Catalog # BOCH10120
Supplier:   BOCHEM
Wire baskets, rectangular, with handle
Catalog # BOCH10120
Supplier:  BOCHEM
Wire baskets, rectangular, with handle
Wire baskets, rectangular, with handle
300×400×200 mm
Price
6 700,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
These rectangular stainless steel baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.
  • Spot-welded grid 8×8×1 mm
  • Reinforced edges
  • Two cross-ties
Specifications
  • Š×H×V:
    300×400×200 mm