Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Trojnožky
Příslušenství pro stativy, RSG solingen
Příslušenství pro stativy, RSG solingen
Příslušenství pro stativy, RSG solingen
Catalog # RSGA497.370
Supplier:
RSG
Expand image
Příslušenství pro stativy, RSG solingen
Catalog # RSGA497.370
Supplier:
RSG
Specifications
Popis:
Tripod extensions, set of three, total height up to 370 mm
Bal.:
3
Specifications
About this item
Product Details
Product Details