Specifications
- Popis:iBox® Scientia 615 system (for 5 mice), motorised fixed lens (25 mm, f/0,95)
- Sekvence:03
- Bal.:1
Specifications
About this item
iBox® Scientia small animal imaging systems are designed to automate in vivo small animal imaging research with one-touch, preset or user-defined PC controls for accurate, repeatable imaging. The iBox® Scientia systems incorporate a fully motorised darkroom, high specification camera and lens, a motorised high intensity light source (450 nm to NIR) and a licence fee to cover the small animal imaging technique patented by AntiCancer, Inc.
- Choice of high resolution, high sensitivity CCD cameras for in vivo imaging
- Motorised optics for setting the aperture, zoom and focus
- Light-tight darkroom creates optimum imaging conditions, with viewing window to enable quick sample inspection
- Matched emission/excitation filter sets specifically for small animal imaging
- Automated control of the five-position emission filter array to allow addition of emissions for visible, NIR and UV to meet all spectral requirements
- Roll-out tray provides easy access for placement of mice on the warming plate
The system is intended for users imaging fluorescence in small animals for many applications, including in vivo tumour growth, stem cell biology, infectious disease, drug efficacy testing, neurobiology, angiogenesis and signal transduction. It is also suitable for busy multi-user laboratories that may have gel imaging or other imaging applications too.
IQ/OQ documentation can be purchased for any iBox® system, if required. Please enquire for further details.