0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Uzávěry
Uzávěry k láhvím
Screw caps, PP, white, with PTFE-Insert and tamper evident ring
Screw caps, PP, white, with PTFE-Insert and tamper evident ring
215-2883
:
Kautex
Screw caps, PP, white, with PTFE-Insert and tamper evident ring
215-2883
:
Kautex
Závit:
32 mm
Popis:
Screw closure PP, white, with tamper evident ring and PTFE cup seal
Bal.:
1.200
Pro láhve:
100 - 250 ml