Beta radiation shields
305×9×480 mm, Angled
Katalogové číslo KART572
Dodavatel: KARTELL
Přejít na:
Product Details & Documents
Acrylic protection shield, transparent, thickness: 9 mm.
- Provides protection from radiations while using beta emitting isotopes, including P32
- Free standing, porable and stable, fitted with non skid feet
Specifikace
- Tvar:Angled
- Š×H×V:305×9×480 mm