Magnetic stirring bars, triangular
Dodavatel: KARTELL
Přejít na:
O tomto produktu
PTFE coated, triangular magnetic stirring bars.
- Scraping action ideal for dissolving solids and mixing sediments
- Angled surfaces create high turbulence even at low speeds
- Suitable for food stuff
4 Dostupných variant níže
Product Details & Documents
PTFE coated, triangular magnetic stirring bars.
- Scraping action ideal for dissolving solids and mixing sediments
- Angled surfaces create high turbulence even at low speeds
- Suitable for food stuff
Product Family Options
Informace o produktu
- SekvenceDélkaØDostupnostPrice