ContactUsLinkComponent
Eyewash bottle
Eyewash bottle
Katalogové číslo 215-1363
Dodavatel:  KARTELL
undefined
Eyewash bottle
Katalogové číslo 215-1363
Dodavatel:  KARTELL
Specifikace
  • Popis:
    Empty eyewash bottle, PE, 500 ml, with specially designed snap-on cap
  • Bal.:
    1
Specifikace
O tomto produktu
This eyewash bottle can be filled with distilled water or other medicated liquid for ophthalmic use and allows a quick and immediate flushing.
  • Provided with an eye cup with snap on cap
  • Airvent valve in the head assures air refill, thus preventing draw back of contaminated wash into the bottle
  • perforated spray plate in the cup centre breaks up the stream to give the eye a gentle wash
  • Drain tube releases waste