Orders LinkContactUsLinkComponent
Eyewash bottle
Eyewash bottle
Empty eye rinsing bottle for 600 ml content, PE
Catalog # BURK2507-2000
Supplier:   BURKLE
Eyewash bottle
Catalog # BURK2507-2000
Supplier:  BURKLE
Eyewash bottle
Eyewash bottle
Empty eye rinsing bottle for 600 ml content, PE
Price
1 360,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Empty bottle for 600 ml contents, leakproof due to valve and dust protection cap. Instructions printed on (in German), made of transparent PE.
Specifications
  • Popis:
    Empty eye rinsing bottle for 600 ml content, PE
Accessories
Product Information
  • Catalog #
    Popis
    Availability
    Price
  • BURK2507-2001
    Wall fixture for mini eyewash station
    Log in for availability
    1 * 1 KS - 705,00 Kč