Advanced imaging system, BioSpectrum®
Catalog # 733-2019
Supplier: UVP ULTRA VIOLET PRODUCTS
Specifications
- Popis:BioSpectrum® 315, manual lift darkroom, motorised zoom lens (8 - 48 mm, f/1,2)
- Sekvence:01
- Š×H×V:440×425×810 mm
- Kamera:12-bit Gel HR 315, uncooled
- Bal.:1
Specifications
About this item
BioSpectrum® imaging systems are fully automated systems designed for chemiluminescent, bioluminescent, fluorescent, and colorimetric imaging. Units include epi-lighting at 365 nm UV, 460 to 470 nm blue and white light, as well as red, gold and green emission filters to cover all the major gel stains.
- Scientific grade CCD camera mounted in the dark room, easily accessible and protected by the camera cover (cameras are upgradeable)
- Light-tight darkroom with UV protected viewing window allowing visualisation of fluorescent gels without opening the door
- Roll-out transilluminator tray for easy insertion and removal of gels
- Access ports for connecting to external BioLite multi-spectral light source (not supplied) for multiplex image capability
- Resolution is 5,0 megapixel
IQ/OQ documentation can be purchased for any BioSpectrum® system, if required. Please enquire for further details. Please contact your local Avantor sales office for full details of the options and accessories available for this system.
* No excitation filters are included with the BioLite unit. Please ask for details of the available choice.
Delivery: Each system includes 12- or 16-bit depth CCD camera (16-bit file depth), darkroom and five-position emission filter wheel with red, gold and green emission filters. VisionWorks®LS image acquisition and enhancement software is required but must be ordered separately. A FirstLight illuminator or Benchtop transilluminator and software need to be added for a fully functional system. A PC is also required but not supplied.