Precision balances, NewClassic, ML series
METB11124820
- Model:ML303/M
- Miska/velikost plošiny:Ø 120 mm
- Sekvence:23
- Bal.:1
- Čitelnost:0,001 g
- Hmotnostní rozsah:320 g
A cost-effective mobile precision balance for accurate and fast weighing tasks.
- MonoBloc - proven technology for fast and accurate result (except ML802E, ML1502E and ML6001E which have strain gauge technology)
- Internal Adjustment - a simple keystroke adjustment with an internal weight (except models denoted with suffix E which require external calibration)
- SmartTrac; graphical display of how much of the range has been used
- Battery operated or mains (AC) operation for flexibility
- Backlit display
: OIML certified models and a full range of accessories are also available. Please see the website or contact the Avantor Technical Office.