- Popis:G:BOX-F³ gel documentation system, 3,8 MP, with lens feedback
- Sekvence:02
- Š×H×V:570 × 430 × 850 (e×clusief PC)
- Kamera:CCD, 12/16 Bit, 4096/65536 Grey Scale
- Bal.:1
The Syngene G:BOX-F³ gel imaging system is designed for general fluorescence applications, including DNA and protein analysis. The range of applications available can be extended by adding lighting modules*. Driven from a database containing hundreds of application protocols, functional control of the G:BOX-F³ is handled by the GeneSys automatic control software, although full manual control is also possible. It has a 3,8 megapixel camera with motorised (f/1,2) lens , which gives high quality images with undetectable noise.
- CCD camera with 3,8 megapixel resolution and motor driven zoom
- Automated image capture, storage and enhancement via GeneSys control software (full manual control also available, if required)
- Images can be saved as JPEG, TIFF or BMP onto computer via Ethernet or USB interface connections
- Darkroom with large access door and safety interlocks has built-in white light and UV to white light conversion screen
- Maximum sample size 275×205 mm
* The modular construction of the G:BOX-F³ allows the choice of a range of lighting options for both epi- and trans-illumination light applications. Please contact your local Avantor sales office for further information.
Dynamic Range: 3,6 to 4,8.
* The modular construction of the G:BOX-F³ allows the choice of a range of lighting options for both epi- and trans-illumination light applications. Please enquire for further details.