Microcentrifuge tubes, 1,5 ml
KART279
: KARTELL
- Type:Eppendorf type, with flat cap
- Verze:Graduated, conical bottom, non sterile
- Kapacita:1,5 ml
- Bal.:1.000
PP, translucent.
- Very good chemical resistance
- Very good temperature resistance
- Flat cap with writing area
Ideal for samples analysis. Depending on centrifuge being used, these tubes can withstand a centrifugal force of up to 11000×g.