Orders LinkContactUsLinkComponent
Dressing scissors
Dressing scissors
Sharp/blunt, 130 mm
Catalog # USBE4240
Supplier:   USBECK Laborgeräte
Dressing scissors
Catalog # USBE4240
Dressing scissors
Dressing scissors
Sharp/blunt, 130 mm
Price
185,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel dressing scissors with fine-blunt blades.
Specifications
  • Type:
    Sharp/blunt
  • Délka:
    130 mm