Deck kit for small bottle roller culture system, Wheaton®
734-1170
- Popis:Deck kit for small bottle roller culture system
- Bal.:1
Attaches quickly and easily to the small bottle bench top roller culture system. Holds an additional two rollers per kit. These units will accept two additional decks. Kit consists of assembled roller deck, four support posts, drive belt, necessary hardware and assembly instructions.
Weight: 2,3 kg