Pocket refractometer, DEF-Chek
Catalog # 635-0507
Dodavatel: REICHERT
Specifikace
- Model:DEF-Chek
- Popis:Digital tester for urea concentration (%) in Diesel Exhaust Fluid (DEF)
- Sekvence:003
- Bal.:1
Specifikace
O tomto produktu
Selective catalytic reduction systems for diesel trucks and buses requires Diesel Exhaust Fluid (DEF) at an exact concentration of 32,5 % urea.
The DEF-Chek offers quick accurate results.