Mechanical overhead stirrers, RKG-00-Bo, TURBOTRON®, IKA®
Supplier: IKA
Certificates
About this item
Powerful, mechanically controlled agitators for intensive mixing with approval for use in Ex-Zone 1 (2G), temperature class T3 acc. to the ATEX 95 directive.
- Suitable for open or pressureless vessels
- Exchangeable stirring tools
Ordering information: Supplied without cables.
Specifications
- Catalog No:
- IKAAU098950
- IKAAU084463
- IKAAU087228
- Model:
- RKG-00-Bo ATEX
- RKG-00-Bo ATEX
- RKG-00-Bo ATEX
- Maximální viskozita:
- 50000 mPas
- 50000 mPas
- 100000 mPas
- Rozsah rychlosti I:
- 20 - 1200 min⁻¹
- 20 - 1200 min⁻¹
- 20 - 1200 min⁻¹
- Maximální kroutící moment:
- ~300 Ncm at 20 - 600 min⁻¹
~150 Ncm at 1200 min⁻¹ - ~300 Ncm at 20 - 600 min⁻¹
~150 Ncm at 1200 min⁻¹ - ~600 Ncm at 20 - 600 min⁻¹
~330 Ncm at 1200 min⁻¹
- Š×H×V:
- 369×288×537 mm
- 369×288×537 mm
- 196×333×550 mm
- Hmotnost:
- 13,8 kg
- 13,8 kg
- 20,0 kg