Staining jars, DURAN®
Staining dishes acc. to Schiefferdecker, for up to 10 slides, 70×90×40 mm
Catalog # SCOT233160003
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Soda-lime glass.
- For 10 slides
These staining jars are suitable for standard microscope slides (76×26 mm).
Caution: Note: Do not clean staining dishes and jars at temperatures above 60°C (glass corrosion is possible).
Specifications
- Popis:Staining dishes acc. to Schiefferdecker, for up to 10 slides
- Š×H×V:70×90×40 mm