Orders LinkContactUsLinkComponent
Staining jars according to Hellendahl, DURAN®
Staining jars according to Hellendahl, DURAN®
Staining dishes acc. to Hellendahl, for up to 16 microscope slides, 90×40×90 mm
Catalog # SCOT233140001
Supplier:   DWK Life Sciences
Staining jars according to Hellendahl, DURAN®
Catalog # SCOT233140001
Supplier:  DWK Life Sciences
Staining jars according to Hellendahl, DURAN®
Staining jars according to Hellendahl, DURAN®
Staining dishes acc. to Hellendahl, for up to 16 microscope slides, 90×40×90 mm
Price
7 810,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
Soda-Lime glass.
For up to 16 microscope slides (76x26 mm).
Caution: Note: Do not clean staining dishes and jars at temperatures above 60°C (glass corrosion is possible).
Specifications
  • Popis:
    Staining dishes acc. to Hellendahl, for up to 16 microscope slides
  • Š×H×V:
    90×40×90 mm